Agar plant cell culture tested

Supplier: Thermo Scientific Chemicals

Synonyms: Agar Agar

H26724-0E H26724-36 H26724-22
AAH26724-0EEA 1475 USD
AAH26724-0E AAH26724-36 AAH26724-22
Agar plant cell culture tested
Agar

CAS No.: 9002-18-0 Odor: Odorless Merck Reference: 14,184 MDL No.: MFCD00081288

Melting Pt: 90
Storage Temperature: Ambient
MDL Number: MFCD00081288
CAS Number: 9002-18-0
EINECS: 232-658-1
Merck Index: 14,184
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36877cbcfaca6a0ace648ee20ae51b3467f2748c&vid=4295a0dcbc54bbbd374a5fb934297574c6848d32&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR