2-Bromobenzyl mercaptan 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Bromobenzylmercaptan, o-Bromotoluene-α-thiol

H26247-03 H26247-06 H26247-MD
AAH26247-03EA 48.2 USD
AAH26247-03 AAH26247-06 AAH26247-MD
2-Bromobenzyl mercaptan 99%
2-Bromobenzyl mercaptan

Purity: 99% CAS No.: 143888-85-1 Molecular Formula: C7H7BrS Formula Weight: 203.10 Flash Point: 110°(230°F) Refractive Index: 1.6170 Boiling Point: 256-257° UN No.: 2810 MDL No.: MFCD04038925

Formula: C₇H₇BrS
MW: 203.1
Boiling Pt: 256…257 °C
Density: 1.527
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD04038925
CAS Number: 143888-85-1
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR