6-Chloro-m-toluidine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H26209-06
H26209-14
AAH26209-06EA
106
USD
AAH26209-06
AAH26209-14
6-Chloro-m-toluidine 98%
6-Chloro-m-toluidine
Purity: 98%
CAS No.: 95-81-8
Molecular Formula: C7H8ClN
Formula Weight: 141.60
Flash Point: 107°(225°F)
Melting Point: 31-33°
Boiling Point: 228-230°
UN No.: 2239
MDL No.: MFCD00007674
Formula:
C₇H₈ClN Boiling Pt: 228…230 °C Melting Pt: 31…33 °C Flash Pt: 107 °C (225 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007674 CAS Number: 95-81-8 EINECS: 202-454-7 UN: 2239 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...