3-Fluoro-4-nitrobenzoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H26168-03
H26168-06
AAH26168-03EA
20.2
USD
AAH26168-03
AAH26168-06
3-Fluoro-4-nitrobenzoic acid 98+%
3-Fluoro-4-nitrobenzoic acid
Purity: 98+%
CAS No.: 403-21-4
Molecular Formula: C7H4FNO4
Formula Weight: 185.11
Melting Point: 173-176°
MDL No.: MFCD01862092
Formula:
C₇H₄FNO₄ Melting Pt: 173…176 °C Storage Temperature: Ambient |
MDL Number:
MFCD01862092 CAS Number: 403-21-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...