(R)-(+)-2,2'-Diamino-1,1'-binaphthalene 97%

Supplier: Thermo Scientific Chemicals
Warning

H25785-03 H25785-MD
AAH25785-03EA 149 USD
AAH25785-03 AAH25785-MD
(R)-(+)-2,2'-Diamino-1,1'-binaphthalene 97%
(R)-(+)-2,2'-Diamino-1,1'-binaphthalene

Purity: 97% CAS No.: 18741-85-0 Molecular Formula: C20H16N2 Formula Weight: 284.36 Melting Point: 242-243° Beilstein Reference: 5279621 Optical Rotation: +150° (c=1 in pyridine) MDL No.: MFCD00145204

Formula: C₂₀H₁₆N₂
Melting Pt: 242…243 °C
Storage Temperature: Ambient
MDL Number: MFCD00145204
CAS Number: 18741-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR