(R)-(+)-2,2'-Diamino-1,1'-binaphthalene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H25785-03
H25785-MD
AAH25785-03EA
149
USD
AAH25785-03
AAH25785-MD
(R)-(+)-2,2'-Diamino-1,1'-binaphthalene 97%
(R)-(+)-2,2'-Diamino-1,1'-binaphthalene
Purity: 97%
CAS No.: 18741-85-0
Molecular Formula: C20H16N2
Formula Weight: 284.36
Melting Point: 242-243°
Beilstein Reference: 5279621
Optical Rotation: +150° (c=1 in pyridine)
MDL No.: MFCD00145204
Formula:
C₂₀H₁₆N₂ Melting Pt: 242…243 °C Storage Temperature: Ambient |
MDL Number:
MFCD00145204 CAS Number: 18741-85-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...