(S)-(-)-2,2'-Diamino-1,1'-binaphthalene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(S)-1,1'-Binaphthyl-2,2'-diamine, (S)-(-)-BINAM, (S)-(-)-1,1'-Binaphthyl-2,2'-diamine, (S)-(-)-1,1'-Bi(2-naphthylamine), (S)-(-)-2,2'-Diamino-1,1'-binaphthyl
H25784-MD
H25784-03
AAH25784-MDEA
53.5
USD
AAH25784-MD
AAH25784-03
(S)-(-)-2,2'-Diamino-1,1'-binaphthalene 97%
(S)-(-)-2,2'-Diamino-1,1'-binaphthalene
Purity: 97%
CAS No.: 18531-95-8
Molecular Formula: C20H16N2
Formula Weight: 284.36
Melting Point: 236-242°
Beilstein Reference: 2419071
Optical Rotation: -150° (c=1 in pyridine)
MDL No.: MFCD00145204
Formula:
C₂₀H₁₆N₂ MW: 284.362 g/mol Melting Pt: 236…242 °C Storage Temperature: Ambient |
MDL Number:
MFCD00145204 CAS Number: 18531-95-8 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...