6-Chloro-7-deazapurine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25713-03
B25713-06
AAB25713-03EA
208
USD
AAB25713-03
AAB25713-06
6-Chloro-7-deazapurine 98%
6-Chloro-7-deazapurine
Purity: 98%
CAS No.: 3680-69-1
Molecular Formula: C6H4ClN3
Formula Weight: 153.52
Storage Temperature: 2°C to 8°C
Melting Point: 190-192°
MDL No.: MFCD01686865
Formula:
C₆H₄ClN₃ Melting Pt: 190…192 °C Storage Temperature: Ambient |
MDL Number:
MFCD01686865 CAS Number: 3680-69-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...