Dimethyl glutarate 98%
Supplier: Thermo Scientific Chemicals
Purity: 98%
CAS No.: 1119-40-0
Molecular Formula: C7H12O4
Formula Weight: 160.17
Flash Point: 103°(217°F)
Refractive Index: 1.4240
Melting Point: -37°
Boiling Point: 213-215°
Beilstein Reference: 1771444
Merck Reference: 14,4473
MDL No.: MFCD00008468
Formula:
C₇H₁₂O₄ Boiling Pt: 213…215 °C Melting Pt: –37 °C Density: 1.087 Flash Pt: 103 °C(217 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008468 CAS Number: 1119-40-0 EINECS: 214-277-2 Merck Index: 14,4473 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...