2-Chloroethyl methyl ether 98%

Supplier: Thermo Scientific Chemicals
Danger

B24081-14 B24081-36 B24081-22
AAB24081-14EA 44 USD
AAB24081-14 AAB24081-36 AAB24081-22
2-Chloroethyl methyl ether 98%
2-Chloroethyl methyl ether

Purity: 98% CAS No.: 627-42-9 Molecular Formula: C3H7ClO Formula Weight: 94.54 Flash Point: 8°(46°F) Refractive Index: 1.4090 Boiling Point: 89-90° UN No.: 3271 MDL No.: MFCD00000972

Formula: C₃H₇ClO
Boiling Pt: 89…90 °C
Melting Pt: –55…55 °C
Density: 1.04
Flash Pt: 8 °C (46 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000972
CAS Number: 627-42-9
EINECS: 210-999-7
UN: 1992
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR