4-Amino-2,2,6,6-tetramethylpiperidine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24062-14
B24062-06
AAB24062-14EA
193
USD
AAB24062-14
AAB24062-03
AAB24062-06
4-Amino-2,2,6,6-tetramethylpiperidine 98%
4-Amino-2,2,6,6-tetramethylpiperidine
Purity: 98%
CAS No.: 36768-62-4
Molecular Formula: C9H20N2
Formula Weight: 156.27
Flash Point: 75°(167°F)
Melting Point: 16-18°
Boiling Point: 188-189°
MDL No.: MFCD00005984
Formula:
C₉H₂₀N₂ Boiling Pt: 188…189 °C Melting Pt: 16…18 °C Density: 0.912 Flash Pt: 75 °C(167 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005984 CAS Number: 36768-62-4 EINECS: 253-197-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...