4-Amino-2,2,6,6-tetramethylpiperidine 98%

Supplier: Thermo Scientific Chemicals
Warning

B24062-14 B24062-06
AAB24062-14EA 193 USD
AAB24062-14 AAB24062-03 AAB24062-06
4-Amino-2,2,6,6-tetramethylpiperidine 98%
4-Amino-2,2,6,6-tetramethylpiperidine

Purity: 98% CAS No.: 36768-62-4 Molecular Formula: C9H20N2 Formula Weight: 156.27 Flash Point: 75°(167°F) Melting Point: 16-18° Boiling Point: 188-189° MDL No.: MFCD00005984

Formula: C₉H₂₀N₂
Boiling Pt: 188…189 °C
Melting Pt: 16…18 °C
Density: 0.912
Flash Pt: 75 °C(167 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005984
CAS Number: 36768-62-4
EINECS: 253-197-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR