Ethyl hydrogen adipate 98%

Supplier: Thermo Scientific Chemicals

B24006-14 B24006-06
AAB24006-14EA 72.4 USD
AAB24006-14 AAB24006-06
Ethyl hydrogen adipate 98%
Ethyl hydrogen adipate

Purity: 98% CAS No.: 626-86-8 Molecular Formula: HO2C(CH2)4CO2CH2CH3 Formula Weight: 174.20 Flash Point: 110°(230°F) Refractive Index: 1.4390 Melting Point: 28-29° Boiling Point: 179-181° dec. MDL No.: MFCD00004419

Formula: C₈H₁₄O₄
Boiling Pt: 179…181 °C (dec.)
Melting Pt: 28…29 °C
Density: 0.980
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004419
CAS Number: 626-86-8
EINECS: 210-966-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR