Saccharin 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: o-Sulphobenzimide, o-Benzoic sulphimide, o-Benzoic sulfimide, o-Sulfobenzimide

B23938-22 B23938-0E B23938-36
AAB23938-22EA 43.64 USD
AAB23938-22 AAB23938-0E AAB23938-36
Saccharin 98+%
Saccharin

An artificial sweetener which is an inhibitor for phosphotransferase activity


Purity: 98+% CAS No.: 81-07-2 Molecular Formula: C7H5NO3S Formula Weight: 183.18 Melting Point: 227-230° Beilstein Reference: 6888 Merck Reference: 14,8311 MDL No.: MFCD00005866

Formula: C₇H₅NO₃S
MW: 183.19 g/mol
Melting Pt: 227…230 °C
Density: 0.828
Storage Temperature: Ambient
MDL Number: MFCD00005866
CAS Number: 81-07-2
EINECS: 201-321-0
Merck Index: 14,8311

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR