3-Fluoro-p-anisaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Fluoro-4-methoxybenzaldehyde, 3-Fluoro-4-anisaldehyde

B23827-03 B23827-06
AAB23827-03EA 23.9 USD
AAB23827-03 AAB23827-06
3-Fluoro-p-anisaldehyde 98%
3-Fluoro-p-anisaldehyde

Purity: 98% CAS No.: 351-54-2 Molecular Formula: C8H7FO2 Formula Weight: 154.14 Flash Point: 110°(230°F) Melting Point: 34-35° Boiling Point: 129-132°/11mm Beilstein Reference: 1942621 MDL No.: MFCD00003349

Formula: C₈H₇FO₂
MW: 154.14
Boiling Pt: 129…132 °C/11 mm
Melting Pt: 34…35 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003349
CAS Number: 351-54-2
EINECS: 206-514-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR