(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(R)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (R)-(+)-BINAP, (R)-BINAP, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
B23785-03
B23785-06
B23785-MD
AAAB23785-03EA
87.8
USD
AAAB23785-03
AAB23785-06
AAB23785-MD
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 98%
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Purity: 98%
CAS No.: 76189-55-4
Molecular Formula: C44H32P2
Formula Weight: 622.69
Melting Point: 239-241°
Beilstein Reference: 4914063
Merck Reference: 14,1223
Optical Rotation: +230° (c=0.3 in toluene)
MDL No.: MFCD00010805
Formula:
C₄₄H₃₂P₂ Melting Pt: 239…241 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010805 CAS Number: 76189-55-4 Merck Index: 14,1223 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...