3-[(Trifluoromethyl)thio]aniline 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23734-06
B23734-03
AAB23734-06EA
332
USD
AAB23734-06
AAB23734-03
3-[(Trifluoromethyl)thio]aniline 96%
3-[(Trifluoromethyl)thio]aniline
Purity: 96%
CAS No.: 369-68-6
Molecular Formula: C7H6F3NS
Formula Weight: 193.19
Flash Point: 103°(218°F)
Refractive Index: 1.5220
Odor: Unpleasant
Boiling Point: 221-224°
UN No.: 2810
MDL No.: MFCD00190130
Formula:
C₇H₆F₃NS Boiling Pt: 221…224 °C Density: 1.348 Flash Pt: 103 °C(218 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00190130 CAS Number: 369-68-6 EINECS: 206-722-4 UN: 2810 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...