2-(Dimethylamino)-ethanol 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Purity: 99+%
CAS No.: 108-01-0
Molecular Formula: C4H11NO
Formula Weight: 89.14
Flash Point: 40°(104°F)
Refractive Index: 1.4295
Odor: Ammonia-like
Melting Point: -60°
Boiling Point: 134-136°
Beilstein Reference: 1209235
Merck Reference: 14,2843
UN No.: 2051
MDL No.: MFCD00002846
Formula:
C₄H₁₁NO Boiling Pt: 134…136 °C Melting Pt: –60 °C Density: 0.886 Flash Pt: 40 °C (104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002846 CAS Number: 108-01-0 EINECS: 203-542-8 UN: 2051 ADR: 8,II Merck Index: 14,2843 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...