Formaldehyde diethyl acetal 99% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethylal, Diethoxymethane
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 100 ppm
B23134-AE
AAB23134-AEEA
36.85
USD
AAB23134-AE
Formaldehyde diethyl acetal 99% stabilized
Formaldehyde diethyl acetal
Purity: 99%
CAS No.: 462-95-3
Molecular Formula: CH3CH2OCH2OCH2CH3
Formula Weight: 104.15
Flash Point: -5°(23°F)
Physical Form: Liquid
Density: 0.831
Refractive Index: 1.3740
Melting Point: -66°
Boiling Point: 87-88°
Beilstein Reference: 1697253
UN No.: 2373
MDL No.: MFCD00009246
Formula:
C₅H₁₂O₂ MW: 104.15 g/mol Boiling Pt: 87…88 °C Melting Pt: –66 °C Density: 0.831 Flash Pt: –5 °C (23 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009246 CAS Number: 462-95-3 EINECS: 207-330-6 UN: 2373 ADR: 3,II |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...