(Ethyl)triphenylphosphonium bromide 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethyltriphenylphosphonium bromide

B23096-22 B23096-14 B23096-36
AAB23096-22EA 102.82 USD
AAB23096-22 AAB23096-14 AAB23096-36
(Ethyl)triphenylphosphonium bromide 98+%
(Ethyl)triphenylphosphonium bromide

Purity: 98+% CAS No.: 1530-32-1 Molecular Formula: C20H20BrP Formula Weight: 371.26 Flash Point: 200°(392°F) Odor: Odorless Beilstein Reference: 3599630 UN No.: 3077 MDL No.: MFCD00011838

Formula: C₂₀H₂₀BrP
MW: 371.26 g/mol
Melting Pt: 206…210 °C
Flash Pt: >200 °C (392 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011838
CAS Number: 1530-32-1
EINECS: 216-223-3
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR