(±)-3-Amino-1,2-propanediol 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22672-14
B22672-36
B22672-22
AAB22672-14EA
72.98
USD
AAB22672-14
AAB22672-36
AAB22672-22
(±)-3-Amino-1,2-propanediol 97%
(±)-3-Amino-1,2-propanediol
Purity: 97%
CAS No.: 616-30-8
Molecular Formula: C3H9NO2
Formula Weight: 91.11
Flash Point: 186°(366°F)
Refractive Index: 1.4920
Boiling Point: 264-266°/739mm
Beilstein Reference: 1719121
UN No.: 2735
MDL No.: MFCD00008140
Formula:
C₃H₉NO₂ Boiling Pt: 264…266 °C/739 mm Density: 1.186 Flash Pt: 186 °C (366 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008140 CAS Number: 616-30-8 EINECS: 210-475-8 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...