Cholecalciferol (Vitamin D3) 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Purity: 99%
CAS No.: 67-97-0
Molecular Formula: C27H44O
Formula Weight: 384.65
Storage Temperature: 2°C to 8°C
Melting Point: 83-87°
Beilstein Reference: 2339331
Merck Reference: 14,10019
Optical Rotation: +108° (c=0.8 in ethanol)
UN No.: 2811
MDL No.: MFCD00078131
Formula:
C₂₇H₄₄O Melting Pt: 83…87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00078131 CAS Number: 67-97-0 EINECS: 200-673-2 UN: 2811 ADR: "6.1",III Merck Index: 14,10019 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...