(S)-(-)-1,2-Epoxypropane 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(S)-(-)-Methyloxirane, (S)-(-)-Propylene oxide, (S)-(-)-Propene oxide
B22341-06
B22341-14
AAB22341-22EA
1021.52
USD
AAB22341-22
AAB22341-06
AAB22341-14
(S)-(-)-1,2-Epoxypropane 99%
(S)-(-)-1,2-Epoxypropane
Purity: 99%
CAS No.: 16088-62-3
Molecular Formula: C3H6O
Formula Weight: 58.08
Flash Point: -37°(-35°F)
Refractive Index: 1.3670
Odor: Ether-like
Boiling Point: 34-35°
Beilstein Reference: 79765
Merck Reference: 14,7856
Optical Rotation: -14° (neat)
UN No.: 1280
MDL No.: MFCD00064312
Formula:
C₃H₆O Boiling Pt: 34…35 °C Melting Pt: –112 °C Density: 0.829 Flash Pt: –37 °C (–35 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064312 CAS Number: 16088-62-3 EINECS: 240-241-0 UN: 1280 ADR: 3,I Merck Index: 14,7856 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...