Polyethylene glycol 400

Supplier: Thermo Scientific Chemicals

Synonyms: PEG 400

B21992-0B B21992-30
AAB21992-0BEA 135.93 USD
AAB21992-0B AAB21992-30
Polyethylene glycol 400
Polyethylene glycol 400

CAS No.: 25322-68-3 Molecular Formula: H(OCH2CH2)nOH Flash Point: 240°(464°F) Refractive Index: 1.4660 Odor: Odorless pH: 5-7 Melting Point: 4-8° Merck Reference: 14,7568 MDL No.: MFCD01779601

Formula: H(OCH₂CH₂)nOH
Boiling Pt: ? 250 °C (1013 hPa)
Melting Pt: 4…8 °C
Density: 1.126
Flash Pt: 240 °C(464 °F)
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2
Merck Index: 14,7568

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR