Diphenylcyclopropenone, (max. 5% H₂O) 99% (dry weight)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,3-Diphenyl-2-cyclopropen-1-one, 2,3-Diphenylcycloprop-2-en-1-one, 2,3-Diphenyl-2-cyclopropen-1-on, 1,2-Diphenylcyclopropen-3-one
B21954-06
B21954-03
B21954-14
AAB21954-06EA
196
USD
AAB21954-06
AAB21954-03
AAB21954-14
Diphenylcyclopropenone, (max. 5% H₂O) 99% (dry weight)
Diphenylcyclopropenone
Purity: 99%
CAS No.: 886-38-4
Molecular Formula: C15H10O
Formula Weight: 206.24
Melting Point: 119-121°
Beilstein Reference: 608049
Merck Reference: 14,3308
MDL No.: MFCD00001311
Formula:
C₁₅H₁₀O MW: 206.24 g/mol Boiling Pt: 407 Melting Pt: 119…121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001311 CAS Number: 886-38-4 EINECS: 212-948-4 Merck Index: 14,3308 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...