Crystal violet

Supplier: Thermo Scientific Chemicals
Danger

B21932-36 B21932-22 B21932-14
AAB21932-36EA 271.42 USD
AAB21932-36 AAB21932-22 AAB21932-14
Crystal violet
Crystal Violet

CAS No.: 548-62-9 Molecular Formula: C25H30ClN3 Formula Weight: 407.99 Melting Point: ca 215° dec. Beilstein Reference: 4077708 Merck Reference: 14,4395 MDL No.: MFCD00011750

Formula: C₂₅H₃₀ClN₃
Melting Pt: ca 215 °C (dec.)
Density: 1.19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011750
CAS Number: 548-62-9
EINECS: 208-953-6
UN: 3077
ADR: "9",III
Merck Index: 14,4395

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR