Sodium methoxide ∼30% (w/w) in methanol

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium methylate, Sodium methanolate

B21872-AP B21872-0F
AAB21872-APEA 84.59 USD
AAB21872-AP AAB21872-0F
Sodium methoxide ∼30% (w/w) in methanol
Sodium methoxide

CAS No.: 124-41-4 Molecular Formula: CH3ONa Formula Weight: 54.02 Flash Point: 16°(61°F) Odor: Alcohol-like Melting Point: 6° Boiling Point: 93° Beilstein Reference: 3592982 Merck Reference: 14,8643 UN No.: 1289 MDL No.: MFCD00012179

Formula: CH₃NaO
Boiling Pt: 93 °C
Melting Pt: <6 °C
Density: 0.969
Flash Pt: 16 °C(61 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012179
CAS Number: 124-41-4
EINECS: 204-699-5
UN: 1289
ADR: 3,III
Merck Index: 14,8643

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR