Safranine O

Supplier: Thermo Scientific Chemicals
Warning

B21674-09 B21674-18
AAB21674-09EA 35 USD
AAB21674-09 AAB21674-18
Safranine O
Safranine O

Redox and adsorption indicator. Biological stain used in histology, cytology, bacteriology


CAS No.: 477-73-6 Molecular Formula: C20H19ClN4 Formula Weight: 350.85 Odor: Odorless Beilstein Reference: 3924099 MDL No.: MFCD00011759

Formula: C₂₀H₁₉ClN₄
Density: 1.447 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011759
CAS Number: 477-73-6
EINECS: 207-518-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR