tert-Butylchlorodimethylsilane 50% (w/w) in toluene

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: tert-Butyldimethylsilyl Chloride, Chloro-tert-butyldimethylsilane, t-Butyldimethylchlorosilane, tert-Butyldimethylchlorosilane, tert-Butyldimethylsilyl chloride, TBDMS chloride

B21286-18
AAB21286-30EA 357.79 USD
AAB21286-30 AAB21286-18 AAB21286-0B
tert-Butylchlorodimethylsilane 50% (w/w) in toluene
tert-Butylchlorodimethylsilane

CAS No.: 18162-48-6 Molecular Formula: C6H15ClSi Formula Weight: 150.73 Flash Point: 7°(45°F) Odor: Characteristic UN No.: 2924 MDL No.: MFCD00000501

Formula: C₆H₁₅ClSi
Density: 0.87
Flash Pt: 7 °C (45 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000501
CAS Number: 18162-48-6
EINECS: 242-042-4
UN: 2920
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR