6-Thioguanine 98%

Supplier: Thermo Scientific Chemicals
Danger

B21280-03 B21280-06
AAB21280-03EA 85.4 USD
AAB21280-03 AAB21280-06
6-Thioguanine 98%
6-Thioguanine

Purity: 98% CAS No.: 154-42-7 Molecular Formula: C5H5N5S Formula Weight: 167.19 Odor: Odorless Melting Point: 360° Beilstein Reference: 157765 UN No.: 2811 MDL No.: MFCD00233553

Formula: C₅H₅N₅S
Melting Pt: >360 °C
Storage Temperature: Ambient
MDL Number: MFCD00233553
CAS Number: 154-42-7
EINECS: 205-827-2
UN: 2811
ADR: "6.1",III
Merck Index: 14,9337

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR