5,6-Dimethoxyindole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21126-03
AAB21126-03EA
309
USD
AAB21126-03
AAB21126-01
5,6-Dimethoxyindole 98%
5,6-Dimethoxyindole
Purity: 98%
CAS No.: 14430-23-0
Molecular Formula: C10H11NO2
Formula Weight: 177.20
Melting Point: 154-160°
Boiling Point: 198°/8mm
MDL No.: MFCD00005675
Formula:
C₁₀H₁₁NO₂ Boiling Pt: 198 °C/8 mm Melting Pt: 154…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005675 CAS Number: 14430-23-0 EINECS: 238-402-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...