4-Methylumbelliferyl-β-D-galactopyranoside 98%
Supplier: Thermo Scientific Chemicals
Purity: 98%
CAS No.: 6160-78-7
Molecular Formula: C16H18O8
Formula Weight: 338.32
Storage Temperature: -30°C to -10°C
Melting Point: ca 260° dec.
Beilstein Reference: 94673
Optical Rotation: -64° (c=1 in pyridine, 24h)
MDL No.: MFCD00036773
Formula:
C₁₆H₁₈O₈ Melting Pt: ∼ 260 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00036773 CAS Number: 6160-78-7 EINECS: 228-185-5 |
Specification Test Results
Optical rotation | -64° (c=1 in pyridine, 24h) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...