O-(tert-Butyldimethylsilyl)hydroxylamine ≥90%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B21001-03
AAAB21001-01EA
38.56
USD
AAAB21001-01
AAB21001-03
O-(tert-Butyldimethylsilyl)hydroxylamine ≥90%
O-(tert-Butyldimethylsilyl)hydroxylamine
Purity: 90+%
CAS No.: 41879-39-4
Molecular Formula: C6H17NOSi
Formula Weight: 147.29
Melting Point: 62-65°
Boiling Point: 87-90°/40mm
UN No.: 3259
MDL No.: MFCD00054944
Formula:
C₆H₁₇NOSi MW: 147.293 g/mol Boiling Pt: 87…90 °C (40 mmHg) Melting Pt: 62…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00054944 CAS Number: 41879-39-4 UN: 3259 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...