N4-Acetylcytosine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-(2-Oxo-1,2-dihydropyrimidin-4-yl)acetamide
B20851-06
B20851-14
AAB20851-22EA
356.36
USD
AAB20851-22
AAB20851-06
AAB20851-14
N4-Acetylcytosine 98%
N4-Acetylcytosine
Purity: 98%
CAS No.: 14631-20-0
Molecular Formula: C6H7N3O2
Formula Weight: 153.14
Melting Point:
Formula:
C₆H₇N₃O₂ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134466 CAS Number: 14631-20-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...