N4-Acetylcytosine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(2-Oxo-1,2-dihydropyrimidin-4-yl)acetamide

B20851-06 B20851-14
AAB20851-22EA 356.36 USD
AAB20851-22 AAB20851-06 AAB20851-14
N4-Acetylcytosine 98%
N4-Acetylcytosine

Purity: 98% CAS No.: 14631-20-0 Molecular Formula: C6H7N3O2 Formula Weight: 153.14 Melting Point: 300° MDL No.: MFCD00134466

Formula: C₆H₇N₃O₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00134466
CAS Number: 14631-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR