1,2-Epoxycyclopentane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Purity: 97%
CAS No.: 285-67-6
Molecular Formula: C5H8O
Formula Weight: 84.12
Storage Temperature: 2°C to 8°C
Flash Point: 10°(50°F)
Refractive Index: 1.4340
Boiling Point: 101-102°
Beilstein Reference: 102495
UN No.: 1993
MDL No.: MFCD00005161
Formula:
C₅H₈O Boiling Pt: 101…102 °C Density: 0.965 Flash Pt: 10 °C (50 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005161 CAS Number: 285-67-6 EINECS: 206-005-6 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...