1-Bromo-3,4,5-trifluorobenzene 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-Bromo-1,2,3-trifluorobenzene

B20607-03 B20607-06
AAB20607-03EA 45 USD
AAB20607-03 AAB20607-06
1-Bromo-3,4,5-trifluorobenzene 98%
1-Bromo-3,4,5-trifluorobenzene

Purity: 98% CAS No.: 138526-69-9 Molecular Formula: C6H2BrF3 Formula Weight: 210.98 Flash Point: 45°(113°F) Refractive Index: 1.4820 Melting Point: -20° Boiling Point: 140° Beilstein Reference: 7249191 UN No.: 1993 MDL No.: MFCD00070738

Formula: C₆H₂BrF₃
MW: 210.98
Boiling Pt: 140
Melting Pt: -20
Density: 1.767
Flash Pt: 45 °C(113 °F)
Storage Temperature: Ambient
MDL Number: MFCD00070738
CAS Number: 138526-69-9
EINECS: 418-480-9
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR