2,6-Bis(trifluoromethyl)benzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B20486-06 B20486-03
AAB20486-06EA 116 USD
AAB20486-06 AAB20486-03
2,6-Bis(trifluoromethyl)benzoic acid 98%
2,6-Bis(trifluoromethyl)benzoic acid

Purity: 98% CAS No.: 24821-22-5 Molecular Formula: C9H4F6O2 Formula Weight: 258.12 Odor: Odorless Melting Point: 136-140° Beilstein Reference: 2057459 MDL No.: MFCD00000376

Formula: C₉H₄F₆O₂
MW: 258.12
Boiling Pt: 260
Melting Pt: 136…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00000376
CAS Number: 24821-22-5
EINECS: 246-479-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR