(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate

L20131-03 L20131-06
AAAL20131-MDEA 74.55 USD
AAAL20131-MD AAAL20131-03 AAAL20131-06
(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane 97%
(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane

Purity: 97% CAS No.: 113451-59-5 Molecular Formula: C10H18N2O2 Formula Weight: 198.26 Storage Temperature: 2°C to 8°C Melting Point: 74-76° Beilstein Reference: 6460311 Optical Rotation: -44° (c=1 in chloroform) MDL No.: MFCD01569250

Formula: C₁₀H₁₈N₂O₂
Melting Pt: 74…76 °C
Storage Temperature: Ambient
MDL Number: MFCD01569250
CAS Number: 113451-59-5

Order Now

Specification Test Results

Optical rotation -44° (c=1 in chloroform)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR