2,3,5,6-Tetrafluorophenylboronic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(2,3,5,6-Tetrafluorophenyl)metaboric acid, (2,3,5,6-Tetrafluorophenyl)boronic acid, Dihydroxy(2,3,5,6-tetrafluorophenyl)borane, 2,3,5,6-Tetrafluorobenzeneboronic acid
L19826-03
AAAL19826-03EA
283
USD
AAAL19826-03
2,3,5,6-Tetrafluorophenylboronic acid 99%
2,3,5,6-Tetrafluorophenylboronic acid
Purity: 99%
CAS No.: 511295-01-5
Molecular Formula: C6H3BF4O2
Formula Weight: 193.90
Melting Point: ca 140-145°
Beilstein Reference: 9200167
Formula:
C₆H₃BF₄O₂ MW: 193.89 g/mol Melting Pt: ∼ 140…145 °C Storage Temperature: Ambient |
MDL Number:
MFCD04039310 CAS Number: 511295-01-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...