1-Butyl-3-methylimidazolium chloride 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Butyl-1-methyl-1H-imidazol-3-ium chloride

L19749-18 L19749-09
AAAL19749-18EA 173.33 USD
AAAL19749-18 AAAL19749-09
1-Butyl-3-methylimidazolium chloride 96%
1-Butyl-3-methylimidazolium chloride

Purity: 96% CAS No.: 79917-90-1 Molecular Formula: C8H15ClN2 Formula Weight: 174.67 Melting Point: ca 73° MDL No.: MFCD03095425

Formula: C₈H₁₅ClN₂
MW: 174.67 g/mol
Melting Pt: 60
Storage Temperature: Ambient
MDL Number: MFCD03095425
CAS Number: 79917-90-1
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR