3,5-Difluoropyridine-2-carbonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,5-Difluoropicolinonitrile, 2-Cyano-3,5-difluoropyridine

L19724-06
AAAL19724-06EA 130 USD
AAAL19724-06 AAAL19724-03
3,5-Difluoropyridine-2-carbonitrile 98%
3,5-Difluoropyridine-2-carbonitrile

Purity: 98% CAS No.: 298709-29-2 Molecular Formula: C6H2F2N2 Formula Weight: 140.09 Melting Point: 30-34° MDL No.: MFCD03788759

Formula: C₆H₂F₂N₂
Melting Pt: 30…34 °C
Storage Temperature: Ambient
MDL Number: MFCD03788759
CAS Number: 298709-29-2
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR