Benzhydryl isothiocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L19249-03
AAAL19249-03EA
41.5
USD
AAAL19249-03
AAAL19249-06
Benzhydryl isothiocyanate 97%
Benzhydryl isothiocyanate
Purity: 97%
CAS No.: 3550-21-8
Molecular Formula: C14H11NS
Formula Weight: 225.31
Melting Point: 58-60°
Boiling Point: 160-165°/4mm
UN No.: 2811
MDL No.: MFCD00046815
Formula:
C₁₄H₁₁NS Boiling Pt: 160…165 °C/4 mm Melting Pt: 58…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00046815 CAS Number: 3550-21-8 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...