(S)-(-)-4-Fluoro-α-methylbenzylamine 99%, ee 99%, ChiPros®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (S)-1-(4-Fluorophenyl)ethanamine, (S)-(-)-1-(4-Fluorophenyl)ethylamine

L19121-03 L19121-14 L19121-06
AAAL19121-03EA 61.6 USD
AAAL19121-03 AAAL19121-14 AAAL19121-06
(S)-(-)-4-Fluoro-α-methylbenzylamine 99%, ee 99%, ChiPros®
(S)-(-)-4-Fluoro-α-methylbenzylamine

Purity: 99% CAS No.: 66399-30-2 Molecular Formula: C8H10FN Formula Weight: 139.17 Flash Point: 68°(154°F) Refractive Index: 1.5020 Melting Point: -30° Boiling Point: 76°/22mm Beilstein Reference: 6791614 Optical Rotation: -33° (neat) UN No.: 2735 MDL No.: MFCD03092999

Formula: C₈H₁₀FN
Boiling Pt: 76 °C/22 mm
Melting Pt: –30 °C
Density: 1.030
Flash Pt: 68 °C(154 °F)
Storage Temperature: Ambient
MDL Number: MFCD03092999
CAS Number: 66399-30-2
EINECS: 679-261-3
UN: 2735
ADR: "8",III

Order Now

Specification Test Results

Optical rotation -33° (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR