Benzenesulfonic acid ∼75% (w/w) in aqueous solution, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
L19077-AU
L19077-AK
L19077-0J
AAAL19077-AUEA
42.5
USD
AAAL19077-AU
AAAL19077-AK
AAAL19077-0J
Benzenesulfonic acid ∼75% (w/w) in aqueous solution, Technical Grade
Benzenesulphonic acid
CAS No.: 98-11-3
Molecular Formula: C6H6O3S
Formula Weight: 158.18
Flash Point:
Formula:
C₆H₆O₃S MW: 158.18 g/mol Boiling Pt: 319 Melting Pt: 50 Density: 1.30 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011689 CAS Number: 98-11-3 EINECS: 202-638-7 UN: 2586 ADR: 8 III Merck Index: 14,1070 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...