5-Methoxy-1H-indole-3-carbaldehyde 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19072-06
L19072-03
AAAL19072-06EA
295
USD
AAAL19072-06
AAAL19072-03
AAAL19072-MD
5-Methoxy-1H-indole-3-carbaldehyde 99%
5-Methoxy-1H-indole-3-carbaldehyde
Purity: 99%
CAS No.: 10601-19-1
Molecular Formula: C10H9NO2
Formula Weight: 175.19
Melting Point: 179-183°
Beilstein Reference: 132769
MDL No.: MFCD00005623
Formula:
C10H9NO2 Melting Pt: 179…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005623 CAS Number: 10601-19-1 EINECS: 234-220-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...