Ammonium hydrogen difluoride 95% remainder mainly ammonium fluoride
Supplier: Thermo Scientific Chemicals
|
Danger
|
L17237-0E
L17237-22
L17237-36
AAAL17237-0EEA
181
USD
AAAL17237-0E
AAAL17237-22
AAAL17237-36
Ammonium hydrogen difluoride 95% remainder mainly ammonium fluoride
Ammonium hydrogen difluoride
Purity: 95%
CAS No.: 1341-49-7
Molecular Formula: H5F2N
Formula Weight: 57.04
Odor: Ammonia-like
pH: 2-3
Melting Point: 120-122°
Boiling Point: 238° dec.
Merck Reference: 14,498
UN No.: 1727
MDL No.: MFCD00012018
Formula:
NH₄HF₂ Boiling Pt: 238 °C (dec.) Melting Pt: 120…122 °C Density: 1.500 Storage Temperature: Ambient |
MDL Number:
MFCD00012018 CAS Number: 1341-49-7 EINECS: 215-676-4 UN: 1727 ADR: 8,II Merck Index: 14,498 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...