3-Methoxybenzenesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Methoxybenzene-1-sulfonyl chloride, 3-Methoxybenzene-1-sulphonyl chloride

L17050-03 L17050-06
AAAL17050-03EA 84.1 USD
AAAL17050-03 AAAL17050-06
3-Methoxybenzenesulfonyl chloride 97%
3-Methoxybenzenesulphonyl chloride

Purity: 97% CAS No.: 10130-74-2 Molecular Formula: C7H7ClO3S Formula Weight: 206.65 Flash Point: 110°(230°F) Refractive Index: 1.5570 Boiling Point: 158-159°/20mm Beilstein Reference: 2093871 UN No.: 3265 MDL No.: MFCD01318155

Formula: C₇H₇ClO₃S
MW: 206.65 g/mol
Boiling Pt: 158…159 °C/20 mm
Density: 1.460
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01318155
CAS Number: 10130-74-2
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR