Silicon dioxide 99.9%, powder 1,5 µm

Supplier: Thermo Scientific Chemicals

Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide

L16987-14 L16987-22
AAAL16987-14EA 78.89 USD
AAAL16987-14 AAAL16987-22
Silicon dioxide 99.9%, powder 1,5 µm
Silicium dioxide

Purity: 99.9% CAS No.: 7631-86-9 Molecular Formula: SiO2 Formula Weight: 60.09 Odor: Odorless Melting Point: 1710° Boiling Point: 2230° Merck Reference: 14,8493 MDL No.: MFCD00011232

Formula: SiO₂
Boiling Pt: 2230 °C
Melting Pt: 1710 °C
Density: 2.0
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 7631-86-9
EINECS: 231-545-4
Merck Index: 14,8493

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR