Bis(pinacolato)diboron 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
4,4,4',4',5,5,5',5'-Octamethyl-2,2'-bi(1,3,2-dioxaborolane), 4,4,5,5-tetramethyl-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane
L16088-06
L16088-14
L16088-03
AAAL16088-06EA
119
USD
AAAL16088-06
AAAL16088-14
AAAL16088-03
Bis(pinacolato)diboron 98+%
Bis(pinacolato)diboron
Purity: 98+%
CAS No.: 73183-34-3
Molecular Formula: C12H24B2O4
Formula Weight: 253.94
Storage Temperature: 2°C to 8°C
Melting Point: 133-139°
Beilstein Reference: 7703552
MDL No.: MFCD00799570
Formula:
C₁₂H₂₄B₂O₄ MW: 253.94 g/mol Melting Pt: 133…139 °C Storage Temperature: Ambient |
MDL Number:
MFCD00799570 CAS Number: 73183-34-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...